Learn More
IGF1R Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IGF1R.
Supplier: Abnova Corporation PAB18506
Description
This receptor binds insulin-like growth factor with a high affinity. It has tyrosine kinase activity. The insulin-like growth factor I receptor plays a critical role in transformation events. Cleavage of the precursor generates alpha and beta subunits. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival. [provided by RefSeq
Specifications
IGF1R | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
IGF1R | |
IGF1R | |
A synthetic peptide corresponding to residues surrounding Y1346 of human IGF1R. | |
100 μg | |
Primary | |
This antibody is specific to IGF1R. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of IGF1R. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216 | |
Rabbit | |
Affinity purification | |
RUO | |
3480 | |
Human, Mouse, Rat | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only