Learn More
IL18 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL18.
Manufacturer: Abnova Corporation PAB18297
Description
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. [provided by RefSeq
Specifications
IL18 | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
IL18 | |
IL18 | |
A synthetic peptide corresponding to amino acids at C-terminus of human IL18. | |
100 μg | |
Primary | |
Human | |
IgG |
Western Blot | |
0.5 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of IL18. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
IGIF/IL-18/IL-1g/IL1F4/MGC12320 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
3606 | |
Lyophilized |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only