Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18BP Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of IL18BP.

Manufacturer:  Abnova Corporation PAB12552

Catalog No. 89-115-919


Add to cart

Description

Description

The protein encoded by this gene is an inhibitor of the proinflammatory cytokine IL18. This protein binds to IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production. This protein is constitutively expressed and secreted in mononuclear cells. The expression of this protein can be enhanced by IFN-gamma. An elevated level of this protein is detected in the intestinal tissues of patients with Crohn's disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: STKDPCPSQPPVC
Specifications

Specifications

IL18BP
Unconjugated
The optimal working dilution should be determined by the end user.
IL18BP
IL18BPa
Rabbit
100 ug
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of IL18BP.
In PBS
Liquid
IL18BP
A synthetic peptide corresponding to amino acids 44-55 of human IL18BP.
RUO
Primary
10068
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only