Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL32 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant IL32.

Manufacturer:  Abnova Corporation PAB16188

Catalog No. 89-118-078


Add to cart

Description

Description

This gene encodes a member of the cytokine family. The protein contains a tyrosine sulfation site, 3 potential N-myristoylation sites, multiple putative phosphorylation sites, and an RGD cell-attachment sequence. Expression of this protein is increased after the activation of T-cells by mitogens or the activation of NK cells by IL-2. This protein induces the production of TNFalpha from macrophage cells. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Specifications

Specifications

IL32
Unconjugated
ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
IL32
IL-32alpha|IL-32beta|IL-32delta|IL-32gamma|NK4|TAIF|TAIFa|TAIFb|TAIFc|TAIFd
Rabbit
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human
ELISA
Rabbit polyclonal antibody raised against recombinant IL32.
Lyophilized from PBS.
Lyophilized
IL32
Recombinant protein corresponding to human IL32.
RUO
Primary
9235
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only