Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-iNOS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1506060.25ML

Catalog No. NBP15060603


Add to cart

Description

Description

iNOS Polyclonal antibody specifically detects iNOS in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

iNOS
Unconjugated
10mM PBS (pH 7.4) and BSA with Sodium Azide
EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II
Rabbit
IgG
0.25 ml
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission
Primary
4843
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Affinity Purified
NOS2
A synthetic peptide derived from the C-terminus of mouse iNOS and nNOS.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Bovine, Human, Mouse, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit