Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSC, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant INSC.

Manufacturer:  Abnova Corporation PAB23449

Catalog No. 89-125-151


Add to cart

Description

Description

In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization (Izaki et al., 2006 [PubMed 16458856]).[supplied by OMIM

Sequence: CLQVENEHVLKSMKACVSETLSMLGQHFGQLLELALTREVQALVRKIDASDNIYTTESTTGNLFSLTQEGAPLCRIIAKEGGVVALFKVCRQDSFRCLY
Specifications

Specifications

INSC
Unconjugated
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
INSC
INSC
Recombinant protein corresponding to amino acids of human INSC.
Antigen affinity purification
RUO
Primary
387755
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against recombinant INSC.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Liquid
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only