Learn More
IRF3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRF3.
Manufacturer:  Abnova Corporation PAB13123
Catalog No. 89-116-376
Add to cart
Description
Description
IRF3 encodes interferon regulatory factor 3, a member of the interferon regulatory transcription factor (IRF) family. IRF3 is found in an inactive cytoplasmic form that upon serine/threonine phosphorylation forms a complex with CREBBP. This complex translocates to the nucleus and activates the transcription of interferons alpha and beta, as well as other interferon-induced genes. [provided by RefSeq
Specifications
Specifications
IRF3
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
IRF3
IRF3
A synthetic peptide corresponding to internal region 14 amino acids of human IRF3.
RUO
Primary
3661
Immunocytochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of IRF3.
In PBS (0.02% sodium azide)
Liquid
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human