Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of IRS1.
Manufacturer: Abnova Corporation PAB26726
Description
Sequence: A-T-Sp-P-ASpecifications
IRS1 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of IRS1. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
Liquid | |
IRS1 | |
A synthetic peptide corresponding to residues surrounding S312 of human IRS1. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine, Rat |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
IRS1 | |
HIRS-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Primary | |
3667 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only