Learn More
ITPKA Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITPKA.
Supplier:  Abnova Corporation PAB4052
Catalog No. 89-131-252
Add to Cart
Description
Description
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro. ITPKA and ITPKB are 68% identical in the C-terminus region. [provided by RefSeq
Specifications
Specifications
ITPKA
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of ITPKA.
In PBS (0.09% sodium azide)
IP3KA
Rabbit
Protein G purification
RUO
3706
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user.
ITPKA
ITPKA
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ITPKA.
400 μL
Primary
Human, Mouse
Liquid
Product Suggestions
Product Suggestions