Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-JMJD2D, PE, Polyclonal Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103357PE
Description
JMJD2D Polyclonal specifically detects JMJD2D in Human samples. It is validated for ICC/IF (Negative).Specifications
JMJD2D | |
Polyclonal | |
ICC/IF (Negative) | |
EC 1.14.11, EC 1.14.11.-, FLJ10251, JHDM3D, JmjC domain-containing histone demethylation protein 3D, JMJD2Dlysine-specific demethylase 4D, jumonji domain containing 2D, Jumonji domain-containing protein 2D, lysine (K)-specific demethylase 4D, MGC141909 | |
Rabbit | |
55 kDa | |
0.1 mL | |
Transcription Factors and Regulators | |
55693 | |
Store at 4C in the dark. |
Immunocytochemistry, ChIP assay | |
PE | |
PBS with 0.05% Sodium Azide | |
KDM4D | |
Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot# Q6B0I6] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only