Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JPH3 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Jph3.
Supplier: Abnova Corporation PAB2555
Specifications
JPH3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Jph3. | |
In PBS (0.09% sodium azide) | |
JP-3/Jp3/MGC124338/MGC124339/MGC124340 | |
Rabbit | |
Protein G purification | |
RUO | |
57340 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
Jph3 | |
Jph3 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of mouse Jph3. | |
400 μL | |
Primary | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction