Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK2, Rabbit, DyLight 649, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.

Manufacturer:  Abnova Corporation PAB15123

Catalog No. 89-117-456


Add to cart

Description

Description

This gene encodes one of the members of the two-pore-domain background potassium channel protein family. This type of potassium channel is formed by two homodimers that create a channel that leaks potassium out of the cell to control resting membrane potential. The channel can be opened, however, by certain anesthetics, membrane stretching, intracellular acidosis, and heat. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Specifications

KCNK2
DyLight 649
Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user.
KCNK2
K2p2.1|MGC126742|MGC126744|TPKC1|TREK|TREK-1|TREK1|hTREK-1c|hTREK-1e
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
88% sequence identity with rat protein. TREK 1 Please note that it is essential to run against membrane preps. NOT recommended for immunohistochemistry.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.
In 50mM sodium borate
Liquid
KCNK2
A synthetic peptide corresponding to amino acids 1-100 of human KCNK2.
RUO
Primary
3776
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only