Learn More
KDEL, Rabbit, FITC, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
Manufacturer:  Abnova Corporation PAB28976
Catalog No. 89-138-456
Add to cart
Description
Description
Specifications
Specifications
KDEL
FITC
Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to KDEL.
200 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide)
Rabbit
Affinity purification
RUO
Primary
Recognizes KDEL proteins, Hsp90b1, Hspa5, PDI and Calr. It may also see PDIA3 and Erp29.