Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDR Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Kdr.

Manufacturer:  Abnova Corporation PAB12134

Catalog No. 89-115-632


Add to cart

Description

Description

Specifications

Specifications

KDR
Unconjugated
Flow Cytometry (1:100) The optimal working dilution should be determined by the end user.
Kdr
6130401C07|Flk-1|Flk1|Krd-1|Ly73|VEGFR-2|VEGFR2
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody has only been shown to recognize the native form (180-190 KDa protein) of VEGFR-2, known as Flk-1 / KDR / VEGFR 2 (Vascular Endothelial Growth Factor Receptor 2), though it was raised against the precursor form. It has not been characterized against the other forms of VEGFR-2. It has not been tested for cross-reactivity with VEGFR-1.
Flow Cytometry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Kdr.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Liquid
Kdr
A synthetic peptide corresponding to amino acids 1300-1367 of mouse Kdr.
RUO
Primary
16542
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only