Learn More
KHK Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KHK.
Supplier: Abnova Corporation PAB4610
Description
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
KHK | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of KHK. | |
In PBS (0.09% sodium azide) | |
KHK | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human KHK. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
KHK | |
Rabbit | |
Protein G purification | |
RUO | |
3795 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only