Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-KIAA1033, (1123 to 1173), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244291

Catalog No. NBP244291


Add to cart

Description

Description

Specifically detects KIAA1033 in Human, Mouse samples, and it is validated for Immunoprecipitation, Western Blotting

WASHC4 Polyclonal antibody specifically detects WASHC4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

KIAA1033
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Affinity Purified
KIAA1033
The epitope recognized by this antibody maps to a region between residue 1123 to 1173 of human WASH complex subunit 7 using the numbering given in entry Q2M389.2 (GeneID 23325).
Immunogen affinity purified
RUO
Primary
23325
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
KIAA1033, Strumpellin And WASH-Interacting Protein, SWIP, WASH Complex Subunit 7
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only