Learn More
KRT19 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KRT19.
Manufacturer: Abnova Corporation PAB18290
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. Unlike its related family members, this smallest known acidic cytokeratin is not paired with a basic cytokeratin in epithelial cells. It is specifically expressed in the periderm, the transiently superficial layer that envelopes the developing epidermis. The type I cytokeratins are clustered in a region of chromosome 17q12-q21. [provided by RefSeq
Specifications
KRT19 | |
Polyclonal | |
Unconjugated | |
Western Blot (0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(0.5 ug/mL) Immunocytochemistry (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
KRT19 | |
KRT19 | |
A synthetic peptide corresponding to human KRT19. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of KRT19. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
CK19/K19/K1CS/MGC15366 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
3880 | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only