Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KT3 tag, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptides of KT3 tag.

Manufacturer:  Abnova Corporation PAB17066

Catalog No. 89-138-277

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Sequence: KPPTPPPEPET
Specifications

Specifications

KT3 tag
Unconjugated
ELISA (2-10 ug/mL) Immunohistochemistry (0.5-5 ug/mL) Immunocytochemistry (0.5-5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide (conjugated with KLH) corresponding to KT3 tag.
RUO
Primary
Not Provided
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptides of KT3 tag.
In PBS
Rabbit
1 mg
Store at 4°C.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only