Learn More
L-asparaginase, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of L-asparaginase.
Supplier:  Abnova Corporation PAB5959
Catalog No. 89-132-043
Add to Cart
Description
Description
Specifications
Specifications
L-asparaginase
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of L-asparaginase.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
LOC547703
A synthetic peptide corresponding to N-terminus of soybean L-asparaginase.
RUO
547703
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
LOC547703
Rabbit
100 μg
Primary
Plant
Liquid
Recommendations
Recommendations