Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant of LAT2.

Manufacturer:  Abnova Corporation PAB1261

Catalog No. 89-115-944


Add to cart

Description

Description

This gene is one of the contiguous genes at 7q11.23 commonly deleted in Williams syndrome, a multisystem developmental disorder. This gene consists of at least 14 exons, and its alternative splicing generates 3 transcript variants, all encoding the same protein. [provided by RefSeq

Specifications

Specifications

LAT2
Unconjugated
The optimal working dilution should be determined by the end user.
LAT2
HSPC046|LAB|NTAL|WBSCR15|WBSCR5|WSCR5
Rabbit
100 ug
Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes the defined epitope of human Non-T cell Activation Linker (NTAL).
Western Blot
Rabbit polyclonal antibody raised against partial recombinant of LAT2.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
LAT2
Recombinant protein corresponding to amino acids 91-243 of human LAT2.
RUO
Primary
7462
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only