Learn More
LCK Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LCK.
Manufacturer: Abnova Corporation PAB18185
Description
This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein is a key signaling molecule in the selection and maturation of developing T-cells. It contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to the plasma membrane and pericentrosomal vesicles, and binds to cell surface receptors, including CD4 and CD8, and other signaling molecules. Multiple alternatively spliced variants, encoding the same protein, have been described. [provided by RefSeq
Specifications
LCK | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
LCK | |
LCK | |
A synthetic peptide corresponding to human LCK. | |
100 μg | |
Primary | |
This antibody is specific to LCK. | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of LCK. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
YT16/p56lck/pp58lck | |
Rabbit | |
Affinity purification | |
RUO | |
3932 | |
Human, Murine, Rat |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only