Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEMD3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of LEMD3.
Manufacturer: Abnova Corporation PAB25750
Description
The MAN antigens, such as MAN1, are nuclear membrane proteins that are recognized by autoantibodies from a patient with a collagen vascular disease. Because MAN1 contains an LEM domain, the gene is also called LEMD3 (for 'LEM domain-containing-3').[supplied by OMIM
Specifications
LEMD3 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of LEMD3. | |
In PBS (0.02% sodium azide) | |
Liquid | |
LEMD3 | |
A synthetic peptide corresponding to 18 amino acids at C-terminus of human LEMD3. | |
Peptide affinity purification | |
RUO | |
Primary | |
23592 | |
Human, Murine |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
LEMD3 | |
MAN1 | |
Rabbit | |
IgG | |
100 ug | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
At least three isoforms of LEMD3 are known to exist; this antibody will detect all isoforms. LEMD3 antibody is predicted to not cross-react with LEMD1 and LEMD2. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only