Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEP Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a recombinant Lep.

Supplier:  Abnova Corporation PAB5111

Catalog No. 89-131-752


Add to Cart

Description

Description

Specifications

Specifications

LEP
Polyclonal
Rabbit polyclonal antibody raised against a recombinant Lep.
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
ob/obese
Rabbit
150 μg
Primary
Mouse
Liquid
ELISA, Immunoprecipitation, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Lep
Lep
Recombinant protein corresponding to mouse Lep.
RUO
16846
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only