Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHB Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against LHB.

Manufacturer:  Abnova Corporation PAB14524

Catalog No. 89-117-116


Add to cart

Description

Description

This gene is a member of the glycoprotein hormone beta chain family and encodes the beta subunit of luteinizing hormone (LH). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism. [provided by RefSeq

Specifications

Specifications

LHB
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Immunohistochemistry (Frozen sections) (1:50-1:200) The optimal working dilution should be determined by the end user.
LHB
CGB4|LSH-B|hLHB
Rabbit
500 uL
Store at 4°C.
Polyclonal
Human
Immunocytochemistry, Immunohistochemistry
Rabbit polyclonal antibody raised against LHB.
In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (1% BSA, 0.05% sodium azide)
Liquid
LHB
Human LHB.
RUO
Primary
3972
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only