Learn More
LLGL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LLGL2.
Supplier:  Abnova Corporation PAB4249
Catalog No. 89-131-335
Add to Cart
Description
Description
The lethal (2) giant larvae protein of Drosophila plays a role in asymmetric cell division, epithelial cell polarity, and cell migration. This human gene encodes a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Specifications
Specifications
LLGL2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of LLGL2.
In PBS (0.09% sodium azide)
HGL/LGL2
Rabbit
Ammonium sulfate precipitation
RUO
3993
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
LLGL2
LLGL2
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human LLGL2.
400 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions