Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243538
Description
Specifically detects LMO4 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
LMO4 Polyclonal specifically detects LMO4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LMO4 | |
Polyclonal | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Breast tumor autoantigen, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 | |
Rabbit | |
18 kDa | |
0.1 mL | |
Cancer, Tumor Suppressors | |
8543 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
LMO4 | |
Full length human LMO4 Recombinant protein. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction