Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-LMW E. coli Host Cell Protein, (72.5mg/mL), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244222

Catalog No. NBP244222


Add to cart

Description

Description

Specifically detects LMW E. coli Host Cell Protein in E. coli samples, and it is validated for ELISA, Western Blotting

LMW E.Coli Host Cell Protein Polyclonal antibody specifically detects LMW E.Coli Host Cell Protein in E. coli samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

LMW E.Coli Host Cell Protein
72.5 mg/mL
Western Blot 1:500 - 1:1000, ELISA 1:1000-1:2000
Delipidation and Defibrination
Anti-LMW E.Coli Host Cell Proteins Antibody was prepared by repeated immunizations with a mixture low molecular weight (LMW) proteins from E.Coli (BL21).
Delipidation and Defibrination
RUO
Primary
E. coli
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Rabbit
Serum
0.1 ml
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only