Learn More
Rabbit anti-LOC727787, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242882
Catalog No. NBP242882
Add to cart
Description
Description
Specifically detects LOC727787 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
KIR3DL2/CD158k Polyclonal antibody specifically detects KIR3DL2/CD158k in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
LOC727787
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Western Blot, Immunocytochemistry, Immunofluorescence
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human LOC727787. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
Human