Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-LPAR1/LPA1/EDG-2, HRP, Polyclonal Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103363H
Description
LPAR1/LPA1/EDG-2 Polyclonal specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
LPAR1/LPA1/EDG-2 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
Rabbit | |
43 kDa | |
0.1 mL | |
Apoptosis, GPCR, Signal Transduction | |
1902 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HRP | |
PBS with No Preservative | |
LPAR1 | |
Synthetic peptide made to an internal portion of human EDG2 (within residues 200-300). [Swiss-Prot# Q92633] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat, Chicken | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only