Learn More
LPL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LPL.
Manufacturer:  Abnova Corporation PAB13684
Catalog No. 89-116-799
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism. [provided by RefSeq
Specifications
Specifications
LPL
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
LPL
HDLCQ11|LIPD
Rabbit
IgG
100 ug
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL
Polyclonal
Different to the related rat sequence by two amino acids.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of LPL.
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Lyophilized
LPL
A synthetic peptide corresponding to amino acids at C-terminus of human LPL.
Affinity purification
RUO
Primary
4023
Bovine, Human