Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK2 Rabbit anti-Human, Mouse, Pig, Primates, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of LRRK2.

Manufacturer:  Abnova Corporation PAB12393

Catalog No. 89-115-769


Add to cart

Description

Description

This gene is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in this gene have been associated with Parkinson disease-8. [provided by RefSeq

Specifications

Specifications

LRRK2
Unconjugated
The optimal working dilution should be determined by the end user.
LRRK2
AURA17|DARDARIN|PARK8|RIPK7|ROCO2
Rabbit
100 uL
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Porcine, Primate, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of LRRK2.
In Tris-glycine (0.05% sodium azide)
Liquid
LRRK2
A synthetic peptide (conjugated with KLH) corresponding to amino acids 2480-2527 of human LRRK2.
RUO
Primary
120892
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only