Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Luciferase, (90mg/mL), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244165

Catalog No. NBP244165

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifically detects Luciferase in Bacteria samples, and it is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Luciferase
90 mg/mL
Western Blot 1:1000 - 1:5000, ELISA 1:5000 - 1:25000, Immunohistochemistry 1:500 - 1:2500, Immunoprecipitation 1:100
Delipidation and Defibrination
Rabbit
IgG
2 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Bacteria
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Luciferase [Photobacterium fischerii]
Delipidation and Defibrination
RUO
Primary
Reconstitute with 2.0 ml deionized water to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only