Learn More
MAFF, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAFF.
Manufacturer: Abnova Corporation PAB17469
Description
The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that lacks a transactivation domain. It is known to bind the US-2 DNA element in the promoter of the oxytocin receptor (OTR) gene and most likely heterodimerizes with other leucine zipper-containing proteins to enhance expression of the OTR gene during term pregnancy. The encoded protein can also form homodimers, and since it lacks a transactivation domain, the homodimer may act as a repressor of transcription. This gene may also be involved in the cellular stress response. Multiple transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Specifications
MAFF | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MAFF. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
U-MAF | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total MAFF protein. | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
MAFF | |
MAFF | |
A synthetic peptide corresponding to internal of human MAFF. | |
RUO | |
23764 | |
Human, Mouse |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only