Learn More
Rabbit anti-MAGEA4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241318
Catalog No. NBP241318
Add to cart
Description
Description
MAGEA4 Polyclonal antibody specifically detects MAGEA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry.
Specifications
Specifications
MAGEA4
1 mg/mL
Western Blot 1- 2 ug/mL ELISA, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Immunofluorescence 20 ug/mL, Immunocytochemistry
Affinity Purified
MAGEA4
Antibody was raised against a 16 amino acid peptide near the amino terminus of human MAGEA4. The immunogen is located within the first 50 amino acids of MAGEA4.
45 kDa
0.1 mg
Cancer
Primary
4103
Human, Mouse, Rat
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Unconjugated
PBS with 0.02% Sodium Azide
MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4
Rabbit
IgG
Peptide affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
MAGEA4 antibody is human specific. At least four isoforms of MAGEA4 are known to exist.