Learn More
MAP2K3 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP2K3.
Manufacturer: Abnova Corporation PAB19110
Description
The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase is activated by mitogenic and environmental stress, and participates in the MAP kinase-mediated signaling cascade. It phosphorylates and thus activates MAPK14/p38-MAPK. This kinase can be activated by insulin, and is necessary for the expression of glucose transporter. Expression of RAS oncogene is found to result in the accumulation of the active form of this kinase, which thus leads to the constitutive activation of MAPK14, and confers oncogenic transformation of primary cells. The inhibition of this kinase is involved in the pathogenesis of Yersina pseudotuberculosis. Multiple alternatively spliced transcript variants that encode distinct isoforms have been reported for this gene. [provided by RefSeq
Specifications
MAP2K3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MAP2K3. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
MAPKK3/MEK3/MKK3/PRKMK3 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
5606 | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) Immunocytochemistry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
MAP2K3 | |
MAP2K3 | |
A synthetic peptide corresponding to C-terminus of human MAP2K3. | |
100 μg | |
Primary | |
Human, Rat | |
IgG |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only