Learn More
MAP2K4 (phospho S80), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K4.
Manufacturer: Abnova Corporation PAB25413
Description
This gene encodes a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. This kinase is phosphorylated, and thus activated by MAP3K1/MEKK. The knockout studies in mice suggested the roles of this kinase in mediating survival signal in T cell development, as well as in the organogenesis of liver. [provided by RefSeq
Sequence: T-H-Sp-I-ESpecifications
MAP2K4 | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
MAP2K4 | |
JNKK|JNKK1|MAPKK4|MEK4|MKK4|PRKMK4|SEK1|SERK1 | |
Rabbit | |
Affinity purification | |
RUO | |
Primary | |
6416 |
Immunohistochemistry (PFA fixed), Western Blot | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K4. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
Liquid | |
MAP2K4 | |
Synthetic phosphopeptide corresponding to residues surrounding S80 of human MAP2K4. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine, Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only