Learn More
MAP4K2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP4K2.
Supplier:  Abnova Corporation PAB4603
Catalog No. 89-131-488
Add to Cart
Description
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1. [provided by RefSeq
Specifications
Specifications
MAP4K2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of MAP4K2.
In PBS (0.09% sodium azide)
BL44/GCK/RAB8IP
Rabbit
Protein G purification
RUO
5871
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
MAP4K2
MAP4K2
A synthetic peptide (conjugated with KLH) corresponding to internal region of human MAP4K2.
400 μL
Primary
Human, Mouse
Liquid
Recommendations
Recommendations