Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against full-length recombinant MAPK3.

Manufacturer:  Abnova Corporation PAB1236

Catalog No. 89-115-742


Add to cart

Description

Description

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described. [provided by RefSeq

Specifications

Specifications

MAPK3
Unconjugated
Western Blot (1:2000) The optimal working dilution should be determined by the end user.
MAPK3
ERK1|HS44KDAP|HUMKER1A|MGC20180|P44ERK1|P44MAPK|PRKM3
Rabbit
Protein A purification
RUO
Primary
5595
Western Blot
Rabbit polyclonal antibody raised against full length recombinant MAPK3.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
MAPK3
Recombinant His fusion protein corresponding to full length human MAPK3.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only