Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of MARCKS.

Supplier:  Abnova Corporation PAB26789

Catalog No. 89-128-431


Add to Cart

Description

Description

The protein encoded by this gene is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis. [provided by RefSeq

Sequence: K-K-Sp-F-K
Specifications

Specifications

MARCKS
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
MARCKS
MARCKS
A synthetic peptide corresponding to residues surrounding S162 of human MARCKS.
100 μg
Primary
Human, Mouse, Rat
Liquid
Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of MARCKS.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
80K-L/FLJ14368/FLJ90045/MACS/PKCSL/PRKCSL
Rabbit
Affinity chromatography
RUO
4082
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only