Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Matriptase/ST14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP242884
Description
Specifically detects Matriptase/ST14 in Mouse samples, and it is validated for Western Blotting
Matriptase/ST14 Polyclonal specifically detects Matriptase/ST14 in Mouse samples. It is validated for Western Blot.Specifications
Matriptase/ST14 | |
Polyclonal | |
Western Blot 1:500-1:3000 | |
EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
Rabbit | |
95 kDa | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ST14 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse ST14. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
6768 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction