Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of MBD1.

Manufacturer:  Abnova Corporation PAB14126

Catalog No. 89-116-945


Add to cart

Description

Description

DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. Five transcript variants of the MBD1 are generated by alternative splicing resulting in protein isoforms that contain one MBD domain, two to three cysteine-rich (CXXC) domains, and some differences in the COOH terminus. All five transcript variants repress transcription from methylated promoters; in addition, variants with three CXXC domains also repress unmethylated promoter activity. MBD1 and MBD2 map very close to each other on chromosome 18q21. [provided by RefSeq

Specifications

Specifications

MBD1
Unconjugated
ELISA (1:300-1:1000) ChIP (1.7 ug/ChIP) The optimal working dilution should be determined by the end user.
MBD1
CXXC3|PCM1|RFT
Rabbit
50 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ChIP assay, ELISA
Rabbit polyclonal antibody raised against synthetic peptide of MBD1.
In PBS (0.05% sodium azide, 0.05% proclin 300)
Liquid
MBD1
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MBD1.
RUO
Primary
4152
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only