Learn More
MCHR1 Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of MCHR1.
Manufacturer: Abnova Corporation PAB26546
Description
The protein encoded by this gene, a member of the G protein-coupled receptor family 1, is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin. [provided by RefSeq
Specifications
MCHR1 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (22-44 ug/mL) The optimal working dilution should be determined by the end user. | |
MCHR1 | |
GPR24|MCH1R|MGC32129|SLC1 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
2847 | |
Human |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of MCHR1. | |
In PBS (0.09% sodium azide) | |
Liquid | |
MCHR1 | |
A synthetic peptide corresponding to 19 amino acids at N-terminal extracellular domain of human MCHR1. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only