Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFSD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of MFSD2.
Supplier: Abnova Corporation PAB19330
Specifications
MFSD2 | |
Polyclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
MFSD2 | |
MFSD2 | |
A synthetic peptide corresponding to 18 amino acids near C-terminus of human MFSD2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of MFSD2. | |
In PBS (0.02% sodium azide) | |
FLJ14490/FLJ35904/FP1147/HMFN0656/PP10484/RP3-342P20.1 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
84879 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction