Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKI67 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of MKI67.

Manufacturer:  Abnova Corporation PAB12384

Catalog No. 89-115-760


Add to cart

Description

Description

This gene encodes a nuclear protein that is associated with and may be necessary for cellular proliferation. Alternatively spliced transcript variants have been described. A related pseudogene exists on chromosome X. [provided by RefSeq

Specifications

Specifications

MKI67
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:3200) The optimal working dilution should be determined by the end user.
MKI67
KIA|Ki-67
Rabbit
50 uL
Store at 4°C. Do not freeze.
Polyclonal
Human, Murine
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of MKI67.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Liquid
MKI67
A synthetic peptide corresponding to amino acids 2100-2200 of human MKI67.
RUO
Primary
4288
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only