Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLANA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MLANA.
Manufacturer: Abnova Corporation PAB18009
Specifications
MLANA | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant MLANA. | |
In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
MART-1/MART1 | |
Rabbit | |
100 μg | |
Primary | |
Recombinant human MART-1 can be used as a positive control. | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
MLANA | |
MLANA | |
Recombinant protein corresponding to human MLANA. | |
RUO | |
2315 | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only