Learn More
MMP-9 (Gelatinase B) Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MMP-9 (Gelatinase B).
Manufacturer:  Abnova Corporation PAB14912
Catalog No. 89-138-028
Add to cart
Description
Description
Specifications
Specifications
MMP-9 (Gelatinase B)
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to MMP-9 (Gelatinase B).
RUO
Primary
This antibody does not react with other MMPs in IHC.
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of MMP-9 (Gelatinase B).
In PBS (0.09% sodium azide)
Rabbit
1 mL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal