Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP15 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of MMP15.

Manufacturer:  Abnova Corporation PAB27077

Catalog No. 89-128-727


Add to cart

Description

Description

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. [provided by RefSeq

Specifications

Specifications

MMP15
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of MMP15.
In PBS, pH 7.2 (0.05% sodium azide)
Liquid
MMP15
A synthetic peptide corresponding to human MMP15.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
MMP15 polyclonal antibody detects endogenous levels of MMP15 protein.
Western Blot
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
MMP15
MT2-MMP|MTMMP2|SMCP-2
Rabbit
Antigen affinity purification
RUO
Primary
4324
Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only