Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MN1.
Supplier: Abnova Corporation PAB17380
Description
Meningioma 1 (MN1) contains two sets of CAG repeats. It is disrupted by a balanced translocation (4;22) in a meningioma, and its inactivation may contribute to meningioma 32 pathogenesis. [provided by RefSeq
Specifications
MN1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MN1. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
MGCR/MGCR1/MGCR1-PEN/dJ353E16.2 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total MN1 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
MN1 | |
MN1 | |
A synthetic peptide corresponding to internal of human MN1. | |
RUO | |
4330 | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction