Learn More
F(ab')2-Rabbit anti-Mouse IgG, PE, Polyclonal, Secondary Antibody, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Manufacturer:  Invitrogen PA184431
Catalog No. PA184431
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For FACS analysis, use 50ul of the suggested working dilution to label 1x10^6 cells in 100ul. Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
Specifications
Specifications
Mouse IgG
Polyclonal
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4
Rabbit
IgG
100 Tests
DO NOT FREEZE!
Secondary
Murine
Flow Cytometry
PE
Purified
Murine IgG.
Ion-exchange chromatography
RUO
Secondary Antibody
Polyclonal