Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Mouse IgM, Clone: RM109, Secondary Antibody,
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma™ 041647
Description
Specifically detects IgM Clone: RM109 in Mouse samples, and it is validated for ELISA
Specifications
IgM | |
Monoclonal | |
Unconjugated | |
Rabbit | |
Protein A purified | |
RUO | |
Secondary | |
Mouse | |
Purified |
ELISA | |
RM109 | |
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide. | |
Mouse IgM | |
100 μg | |
Secondary, Control Antibodies | |
This antibody reacts to mouse IgM. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, human IgG, rat IgG, or goat IgG. The Fc region of RM109 has been engineered to eliminate the binding of Fc receptor. | |
2°C to 8°C for one year from date of shipment | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction